Learn More
Julabo USA Inc DYNEO DD-BC12.D heating circulator, 115V/60Hz, 20…200C. 1kW heater. 12L tank. CSA/UL
Supplier: Julabo USA Inc 9021512.22.D
Heating circulator, JULABO, DYNEO DD-BC12.D, 115V/60Hz, 20…200C. 1kW heater. Adjustable centrifugal pump 8-27LPM, 1.5-11.6 PSI. USB-A data-logging; USB-B and RS232 external communication. Lemosa Pt100 port for external temperature control and monitoring. Color screen with click and scroll navigation. Programmable. M16x1 pump fittings with 8 and 12mm barbs. 6...7.5L bath tank. Bath tank opening: 8.7x5.9/7.9 in (WxL/D); Dimensions: 13 x 19.3 x 18.5 in. 26.2lbs. CSA/UL marked.