Learn More
Julabo USA Inc DYNEO DD-BC4.D heating circulator, 115V/60Hz, 20…200C. 1kW heater. 4L tank. CSA/UL
Supplier: Julabo USA Inc 9021504.22.D
Heating circulator, JULABO, DYNEO DD-BC4.D, 115V/60Hz, 20…200C. 1kW heater. Adjustable centrifugal pump 8-27LPM, 1.5-11.6 PSI. USB-A data-logging; USB-B and RS232 external communication. Lemosa Pt100 port for external temperature control and monitoring. Color screen with click and scroll navigation. Programmable. M16x1 pump fittings with 8 and 12mm barbs. 3...4.5L bath tank. Bath tank opening: 5.1x5.9/5.9 in (WxL/D); Dimensions: 9.7 x 16.1 x 16.5 in. 18.7lbs. CSA/UL marked.