Learn More
Medchemexpress LLC Dynorphin A (1-10) (TFA) | 98.6% | 1348.48 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP1594A10MG
Encompass_Preferred
Dynorphin A (1-10) (TFA) is an endogenous opioid neuropeptide that binds to extracellular loop 2 of the κ-opioid receptor. It also blocks NMDA-activated current with an IC50 of 42.0 μM. Studies show that Dynorphin A (1-10) (TFA) blocks NMDA-activated currents in a voltage-independent manner.
  • Endogenous opioid neuropeptide
  • Binds to extracellular loop 2 of the κ-opioid receptor
  • Blocks NMDA-activated current
  • IC50 of 42.0 μM for NMDA receptor blockade
  • Voltage-independent action on NMDA receptors
Catalog No. 50-004-22173