Learn More
Carbosynth LLC. Dynorphin A (Human, 1-13) (0.5 mg vial), 1ea, MW: 1604, Molecular formula: C75H126N24O15, CAS# 72957-38-1
SDP
Supplier:  Carbosynth LLC. PDY4080V0.5MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: Tyr-Gly-Gly-Phe-Leu-Arg-Arg-Ile-Arg-Pro-Lys-Leu-Lys, Storage Temperature: -20C, References: A. Goldstein, S. Tachibana, L.I. Lowney, M. Hunkapiller, and L. Hood, Proc. Natl. Acad. Sci. U.S.A., 76, 6666 (1979). (Original; Porcine), S. Horikawa, T. Takai, M. Toyosato, H. Takahashi, M. Noda, H. Kakidani, T. Kubo, T. Hirose, S. Inayama, H. Hayashida, T. Miyata, and S. Numa, Nature, 306, 611 (1983). (Nucleotide Seq.; Human), J. Hughes, Br. Med. Bull., 39, 17 (1983). (Review)
Catalog No. 50-168-6432