Learn More
Cayman Chemical Dynorphin A trifluoroacetate
SDP
Supplier:  Cayman Chemical 18169500 UG
Encompass_Preferred
A heptadecapeptide that acts as an endogenous opioid receptor agonist (Kis = 0.5-1 nM for κ, µ, and δ); has been implicated in antinociceptive functions
Catalog No. 50-296-5002