Learn More
Sino Biological Peptide Substrates: DYRKtide
Supplier:  Sino Biological D9658
Encompass_Preferred
The synthetic peptide (RRRFRPASPLRGPPK) contains a serine/threonine protein kinase phosphorylation site and is routinely evaluated as a substrate for DYRK family kinases.
Catalog No. 50-256-9384