Learn More
Strem, An Ascensus Company CAS# 7429-91-6. 5g. Dysprosium powder (99.9% REO). MFCD00010982
SDP
Supplier:  Strem, An Ascensus Company 9366355G
Encompass
CAS# 7429-91-6. 5g. Dysprosium powder (99.9% REO). MFCD00010982. Molecular Weight: 162.5. Molecular Formula: Dy. Color/form: -40 mesh Strem# 93-6635. http://www.strem.com/catalog/v/93-6635/
Catalog No. 50-901-16131