Learn More
Miele, Inc. E 106/1 Half Insert with spring clips (28 clips 105 mm tall)
Supplier:  Miele, Inc. 03808320
Encompass_Preferred
E 106/1 Mesh Basket
For the optimum loading of wide neck flasks, measuring cylinders, etc.
Equipped with 26 small spring hooks x 105 mm height
The distance between the spring hooks is approx. 60 mm
Visit www.mieleusa.com/professional for technical information.
Catalog No. 50-171-3012