Learn More
Medchemexpress LLC (E)-2-((1,4-dimethylpiperazin-2-ylidene)amino)-5-nitro-N-phenylbenzamide | 1613465-33-0 | 99.7% | 367.40 g/mol | C19H21N5O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY129285MG
Encompass_Preferred
ML336 is a quinazolinone-based small-molecule inhibitor of Venezuelan equine encephalitis virus (VEEV) that potently inhibits VEEV-induced cytopathic effects in multiple strains in the low-nanomolar range. It is supplied for research use only and is typically provided as a powder for biochemical and antiviral assays.
  • Potent VEEV inhibitor exhibiting low-nanomolar antiviral activity.
  • Quinazolinone-based small molecule for antiviral research and probe development.
  • High purity (≈99.65%) for consistent experimental results.
  • Available in small mass vials suitable for screening and assay optimization.
  • Powder stable at -20°C for long-term storage; solutions stable at -80°C.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-62781