Learn More
Medchemexpress LLC (E)-2-(1H-Benzo[d]imidazol-2-ylthio)-N'-(5-bromo-2-hydroxybenzylidene)propanehydrazide | 330676-02-3 | MFCD01230899 | 99.0% | 419.30 g/mol | C17H15BrN4O2S | 25 MG

Supplier: Medchemexpress LLC HY10319425MG
KH7 is a research-grade small-molecule inhibitor of soluble adenylyl cyclase (sAC) used to modulate cAMP signaling in vitro. It is supplied as a solid for laboratory research and should be handled according to standard safety practices.
- Selective soluble adenylyl cyclase inhibitor with reported IC50 of 3-10 μM.
- High purity, typically 99.0%.
- Available in small research pack sizes, including 25 mg.
- Molecular weight 419.30 g/mol; molecular formula C17H15BrN4O2S.
- Soluble in dimethyl sulfoxide (DMSO) for assay preparation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.