Learn More
Medchemexpress LLC (E)-2-(2,5-dimethoxyphenyl)-3-(4-hydroxy-3-methoxyphenyl)-N-[2-(4-hydroxyphenyl)ethyl]prop-2-enamide | 863193-70-8 | 99.6% | 449.50 g·mol-1 | C26H27NO6 | 10MM 1ML

Supplier: Medchemexpress LLC HY12350610MM 1ML
Fenlean is a natural squamosamide-derived Src tyrosine kinase inhibitor intended for research use in studies of microglial activation and neuroprotection. The compound is supplied as solid powder and as a ready-to-use 10 mM solution in DMSO, with documented purity, molecular formula, and storage recommendations to support reproducible in vitro and in vivo experiments.
- High purity for reliable experimental results.
- Available as solid powder and 10 mM solution in DMSO.
- Good solubility in DMSO; in vitro solubility reported up to 100 mg/mL.
- Multiple pack sizes to support screening and scale-up.
- Recommended storage conditions provided to preserve stability.
- CAS, molecular formula, and molecular weight documented for identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.