Learn More
Medchemexpress LLC (E)-2,3',4,5'-tetramethoxystilbene | 24144-92-1 | MFCD03428621 | 99.4% | 300.35 g·mol^-1 | C18H20O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1934010MG
Encompass_Preferred
TMS ((E)-2,3',4,5'-tetramethoxystilbene) is a methylated resveratrol analogue used in research as a selective and competitive inhibitor of cytochrome P450 1B1 (CYP1B1). It shows potent in vitro activity (IC50 ≈ 6 nM, Ki ≈ 3 nM) and is commonly applied in enzymatic assays and cellular studies investigating CYP1B1-related pathways and anti-cancer effects.
  • High purity (99.4%) suitable for biochemical assays.
  • Soluble in DMSO at ≥ 50 mg/mL for in vitro applications.
  • Formulated for in vivo dosing using common vehicle protocols.
  • Multiple pack sizes available to support screening and scale-up studies.
  • Stable under recommended storage conditions for long-term use.
Catalog No. 50-002-95176