Learn More
Medchemexpress LLC (E)-2-cyano-3-[1-[3-(dimethylamino)propyl]indol-3-yl]-N-octylprop-2-enamide | 1128165-88-7 | MFCD19690957 | 99.3% | 408.58 g·mol⁻¹ | C25H36N4O | 1 MG

Supplier: Medchemexpress LLC HY1075451MG
Dynole 34-2 is a small-molecule inhibitor of dynamin GTPases used in cell biology to block receptor-mediated endocytosis and induce apoptosis. It is supplied as a light-yellow solid at high purity for in vitro and in vivo research.
- Potent dynamin GTPase inhibitor (IC50 ≈ 6.9 μM for dynamin-1, 14.2 μM for dynamin-2).
- High purity (99.3%).
- Molecular weight 408.58 g·mol⁻¹.
- Chemical formula C25H36N4O.
- Soluble in DMSO (100 mg/mL); in vivo formulations: 10% DMSO/40% PEG300/5% Tween-80/45% saline or 10% DMSO/90% corn oil.
- Storage: solid at 4°C protected from light; solutions at -80°C up to 6 months or -20°C up to 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.