Learn More
Medchemexpress LLC (E)-2'-deoxy-2'-(fluoromethylene)cytidine | 130306-02-4 | MFCD00893181 | 100.0% | 257.22 | C10H12FN3O4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1060141MG
Encompass_Preferred
Tezacitabine is a nucleoside analogue and ribonucleotide reductase inhibitor used in research on DNA/RNA synthesis and anticancer mechanisms. It is supplied for laboratory use in solid form and as a DMSO solution, with a molecular weight of 257.22 and formula C10H12FN3O4 (CAS 130306-02-4).
  • Ribonucleotide reductase inhibitor for DNA/RNA synthesis studies.
  • Available as solid and as a DMSO solution for flexible dosing.
  • High purity (>99.9%) suitable for research and analytical applications.
  • Stable when stored at -80°C for long-term use.
  • Multiple pack sizes support small-scale and larger experiments.
Catalog No. 50-002-41875