Learn More
Cayman Chemical E-2-Hexadecenal 5mg
SDP
Supplier:  Cayman Chemical 175665 MG
Encompass_Preferred
A sphingolipid degradation product that has been shown to induce cytoskeletal reorganization and eventual apoptosis via a JNK-dependent pathway; reacts readily with deoxyguanosine and DNA to form aldehyde-derived DNA adducts
Catalog No. 50-298-0083