Learn More
Medchemexpress LLC (E)-3-(1H-indol-3-yl-4,5,6,7-d4)acrylic acid | MFCD00005633 | 99.4% | 191.22 g/mol | C11H5D4NO2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW015273S1MG
Encompass_Preferred
3-Indoleacrylic acid-d4 is the deuterium-labeled isotopologue of 3-indoleacrylic acid provided as a high-purity stable-isotope standard for research and analytical use. It is intended for use as an internal standard or tracer in mass spectrometry and metabolic studies, delivering precise isotopic labeling for improved quantitation and identification.
  • Deuterium-labeled (d4) isotopologue for use as a stable-isotope standard.
  • High purity suitable for analytical applications.
  • Accurate mass shift for unambiguous mass spectrometry identification.
  • Available in mg-scale packages for method development and validation.
  • Suitable as an internal standard in quantitative assays.
Catalog No. 50-003-03699