Learn More
Medchemexpress LLC (E)-3-(4-methoxyphenyl)acrylic acid | 943-89-5 | MFCD00004398 | 99.7% | 178.18 g/mol | C10H10O3 | 100 G

Supplier: Medchemexpress LLC HYW068771100G
(E)-3-(4-methoxyphenyl)acrylic acid is a small-molecule endogenous metabolite supplied for research use. It exhibits reported hepatoprotective, anti-amnesic, cognition-enhancing, antihyperglycemic, and neuroprotective activities, and is provided with supporting quality and safety documentation.
- Purity 99.72%.
- CAS number 943-89-5.
- Molecular formula C10H10O3.
- Molecular weight 178.18 g/mol.
- Available as 100 g package.
- Includes data sheet, certificate of analysis, and safety data sheet.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.