Learn More
Medchemexpress LLC (E)-3-pentenoic acid | 1617-32-9 | MFCD00063291 | ≥95.0% | 100.12 g/mol | C5H8O2 | 5 G

Supplier: Medchemexpress LLC HYW0885015G
(E)-3-pentenoic acid is an unsaturated carboxylic acid used as a research reagent and synthetic building block in organic chemistry. It is supplied as a high-purity liquid for laboratory synthesis and analytical applications.
- Unsaturated carboxylic acid used in organic synthesis.
- High purity (≥95.0%).
- Molecular formula C5H8O2 and molecular weight 100.12 g/mol.
- Commonly used for esterification, oxidation, and reduction reactions.
- Available as a 5 g pack for laboratory use.
- For research use only; not intended for human consumption.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.