Learn More
Medchemexpress LLC (E)-3-pyridin-3-yl-1-pyridin-4-ylprop-2-en-1-one | 13309-08-5 | MFCD00224002 | 99.9% | 210.24 g/mol | C13H10N2O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1101695MG
Encompass_Preferred
(E/Z)-3PO is a small-molecule inhibitor of PFKFB3 used to reduce glycolytic flux and investigate metabolic and angiogenesis-related pathways in cellular and biochemical research. It is supplied as an off-white to light yellow solid with high reported purity and is commonly applied in studies of cancer, pulmonary fibrosis, acute lung injury, and atherosclerosis; follow supplier storage recommendations to maintain stability.
  • Potent PFKFB3 inhibitor for glycolysis modulation.
  • Applicable to cellular and biochemical assays.
  • Useful in research on cancer, fibrosis, and inflammatory injury.
  • Off-white to light yellow solid physical form.
  • High reported purity suitable for experimental use.
  • Stable when stored as recommended by the supplier.
  • Molecular formula C13H10N2O and molecular weight ~210.24 g/mol.
Catalog No. 50-002-64809