Learn More
Miele, Inc. E 351 160 mm Injector (Ø 4.0 mm) for use with E 353
Supplier:  Miele, Inc. 03809500
Encompass_Preferred
E 351 Injector nozzle
For the optimum cleaning of the wash items. Size 4 x 160 mm.
Suitable for use in a mobile injector unit
Can be combined with clip E 353
Ø 4 x 160 mm, screw connection
Visit www.mieleusa.com/professional for technical information.
Catalog No. 50-171-3008