Learn More
Medchemexpress LLC (E)-4-(2-(N-((4-methoxyphenyl)sulfonyl)acetamido)styryl)pyridine 1-oxide | 173529-46-9 | 99.9% | 10 MG

Supplier: Medchemexpress LLC HY1204510MG
HMN-214 is an orally bioavailable prodrug of HMN-176 that inhibits polo-like kinase 1 (PLK1) and demonstrates antitumor activity. Supplied as a solid research compound with high purity, defined storage conditions, and recommended solvent for handling.
- Orally bioavailable prodrug with PLK1 inhibitory activity.
- High purity available (99.9%).
- Molecular weight 424.47 and formula C22H20N2O5S.
- Soluble in DMSO (~8.33 mg/mL); ultrasonic assistance recommended.
- Store powder at -20°C for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.