Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-4-(2-Nitrophenyl)-4-oxobut-2-enoic acid | 80937-24-2 | 95.0% | 221.17 | 500 MG
SDP

Supplier:  Medchemexpress LLC HY400142500MG

Encompass_Preferred

(E)-4-(2-Nitrophenyl)-4-oxobut-2-enoic acid is an intermediate that can be used to synthesize Phenyl-4-oxo-2-butenoic acid. Phenyl-4-oxo-2-butenoic acid has high antiulcer and antisecretory activities. It is also an intermediate used in the synthesis of Phenyl-4-oxo-2-butenoic acid, which exhibits high antiulcer and antisecretory activities.

  • Molecular weight: 221.17
  • Formula: C10H7NO5
  • Appearance: Solid
  • Color: Light brown to brown
  • Storage conditions (powder): -20°C for 3 years, 4°C for 2 years
  • Storage conditions (in solvent): -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-03734


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.