Learn More
Medchemexpress LLC (E)-4-hydroxycrotonic acid | 24587-49-3 | MFCD00673880 | 98.0% | 102.09 g/mol | C4H6O3 | 25 MG

Supplier: Medchemexpress LLC HY10098225MG
(E)-4-Hydroxycrotonic acid is a research small molecule used as a high-affinity ligand for gamma-hydroxybutyrate (GHB) binding sites. It is employed in pharmacological studies and for developing three-dimensional pharmacophore models to probe receptor interactions and structure-activity relationships.
- High-affinity ligand for GHB binding sites, useful in receptor studies.
- Purity 98.0% supports reliable experimental results.
- Small molecular weight (102.09 g/mol) facilitates solubility and handling.
- Available in milligram quantities suitable for assay work.
- Storage recommendations support long-term stability of powder and solutions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.