Learn More
Selleck Chemical LLC E-4031 dihydrochloride
SDP
Supplier:  Selleck Chemical LLC S662725MG
Encompass_Preferred
E-4031 is an experimental class III antiarrhythmic drug that blocks potassium channels of the hERG-type
Catalog No. 50-000-02094