Learn More
Cayman Chemical E-5- 2-BromovInyluracIl 250mg
SDP
Supplier:  Cayman Chemical 21030250 MG
Encompass_Preferred
A pyrimidine base; is an inactive metabolite of the antiviral agents sorivudine and BVDU that may be regenerated to BVDU in vivo; irreversibly inactivates DPD in an NADPH-dependent manner; enhances the efficacy of the chemotherapeutic agent and DPD substrate 5-FU in a P388 murine leukemia model at 200 µmol/kg, increasing survival time
Catalog No. 50-298-3831