Learn More
Medchemexpress LLC (E)-5-(dimethylamino)-2-(pyridin-2-yldiazenyl)phenol methanesulfonate | 2756327-67-8 | 95.0% | 338.38 g·mol⁻¹ | C14H18N4O4S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1398935MG
Encompass_Preferred
ERGi-USU-6 mesylate is the methanesulfonate salt of a small-molecule inhibitor that targets the ERG transcription factor and RIOK2 kinase. It has reported in vitro anticancer activity against ERG-positive prostate cancer cell lines and is supplied for laboratory research use only. The material is provided as a solid (brown to black) with reported purity of 95.0% and supporting documentation available.
  • Inhibits ERG transcription factor and RIOK2 kinase.
  • Demonstrates anticancer activity in ERG-positive prostate cancer cell lines.
  • Supplied for research use only; not for human or veterinary use.
  • Provided as a solid, brown to black, with 95.0% purity.
  • Includes datasheet, certificate of analysis, and SDS for quality and safety information.
Catalog No. 50-002-22212