Learn More
Selleck Chemical LLC E-64 10mg 66701-25-5
SDP
Supplier:  Selleck Chemical LLC S737910MG
Encompass_Preferred
E-64 is an irreversible and selective cysteine protease inhibitor, and also inhibits papain, calpain, and cathepsins B and H, but not serine proteases or aspartic proteases. The IC50 for papain is 9 nM. E-64 induces oxidative stress and apoptosis in Filarial Parasite. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4718