Learn More
TARGETMOL CHEMICALS INC E-64 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T603725MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. E-64 (Proteinase inhibitor E 64) is an irreversible and specific cysteine protease inhibitor. The IC50 of E-64 for papain is 9 nM. purity: 99%
Catalog No. 50-257-9624