Learn More
Sigma Aldrich Fine Chemicals Biosciences E-64 protease inhibitor5MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences E31325MG
Encompass
E-64 is a cysteine protease inhibitor that was isolated from the mold Aspergillus japonicus TPR-64. E-64 is also known as N-[N-(L-3-trans-carboxyoxiran-2-carbonyl)-L-leucyl]-agmatine. E-64 effectively inhibits various cysteine proteases in particularbullnbspcathepsin Kbullnbspcathepsin Lbullnbspcathepsin SE-64 also acts against other enzymes such asbullnbspcalpainbullnbspcathepsin Bbullnbspcathepsin Hbullnbsppapain
Catalog No. 50-290-2035