Learn More
Cayman Chemical E-64d, 88321-09-9, 1 mg
SDP
Supplier:  Cayman Chemical 135331
Encompass_Preferred
Molecular Formula C17H30N2O5, Purity ≥98%, Formula Weight 342.4, Synonyms: Aloxistatin, E-64c ethyl ester, EP 453, EST, Loxistatin, NSC 694281; An irreversible, membrane-permeable inhibitor of lysosomal and cytosolic cysteine proteases; at 20-200 M arrests human epidermoid carcinoma A431 cells at mitotic metaphase; inhibits protease-resistant protein accumulation in scrapie-infected neuroblastoma cells (IC50 = 0.5 M).
Catalog No. NC9077982