Learn More
Abcam E-64d (Alo x istatin), cathepsin B and L inhibitor, 1MG
Supplier:  Abcam AB1440481MG
Encompass_Preferred
MW 342.4 Da, Purity >98%. Irreversible cell-permeable cathepsin B and L inhibitor. Inhibits calpain. Inhibits autophagy. Inhibits platelet proteolysis. Reduces brain amyloid-β levels. Shows neuroprotective effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6042