Learn More
Carbosynth LLC. H-Glu[Cyclo(Arg-Gly-Asp-D-Phe-Lys)]2, 25mg, MW: (M.W. 1318.47), Molecular formula: C59H87N19O16
SDP
Supplier:  Carbosynth LLC. PCI3651PI25MG
Encompass_Preferred
Synonyms: H-Glu[cyclo(Arg-Gly-Asp-D-Phe-Lys)]2, E-[c(RGDfK)2], Application: αvß3 Integrin Binding RGD Peptide; RGD Tumor Targeting Peptide (Requires further derivatization before use), Salt Form:, Acetate Salt, Storage Temperature: -20°C, References: M.L. Janssen, W.J. Oyen, I. Dijkgraaf, L.F. Massuger, C. Frielink, D.S. Edwards, M. Rajopadhye, H. Boonstra, F.H. Corstens, and O.C. Boerman, Cancer. Res., 62, 6146 (2002)., Y. Wu, X. Zhang, Z. Xiong, Z. Cheng, D.R. Fisher, S. Liu, S.S. Gambhir, and X. Chen, J. Nucl. Med., 46, 1707 (2005).
Catalog No. 50-168-6271