Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-Cardamonin ((E)-Cardamomin) | 19309-14-9 | 270.28 | 1 ML
SDP

Supplier:  Medchemexpress LLC HYN137810MM/1ML

Encompass_Preferred

(E)-Cardamonin ((E)-Cardamomin) is a novel antagonist of hTRPA1 cation channel with an IC50 of 454 nM.

  • Purity: 99.84%
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Formula: C16H14O4
  • Molecular weight: 270.28
  • Structure classification: Flavonoids, chalcones, phenols, polyphenols
  • Initial source: Plants Zingiberaceae Alpinia katsumadai Hayata
  • Storage: Powder at -20°C for 3 years, 4°C for 2 years; in solvent at -80°C for 2 years, -20°C for 1 year
  • Solubility (in vitro): DMSO at 150 mg/mL (554.98 mM); requires ultrasonic
  • Target: hTRPA1 cation channel with an IC50 of 454 nM

Catalog No. 50-004-14807


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.