Learn More
LGC GENOMICS LLC (Lucigen) E. cloni 10G Chemically Competent Cells, 24 Rxns (DUOs)Clone, construct libraries, transform Gibson Assembly reactions all with high-yield, T1 phage-resistant cells in high efficiencies
Supplier:  LGC GENOMICS LLC 601072
Encompass
Lucigens E. cloni competent cells share the most useful genetic elements of standard cloning strains like DH5™, DH10B™, JM109, TOP10, etc. and directly replace them in cloning protocols. E. cloni chemically competent cells provide the performance researchers need with ease of useonly common laboratory equipment is required. These cells provide solutions for a wide range of applications at economical prices. E. cloni 10G Competent Cells. Routine cloning, subcloning, and plasmid isolation with or without blue/white screening.
Catalog No. 50-104-7942