Learn More
LGC GENOMICS LLC (Lucigen) E. cloni 10G ELITE Electrocompetent Cells, 12 Rxns (DUOs)Clone, construct libraries, transform Gibson Assembly reactions all with high-yield, T1 phage-resistant cells in high efficiencies
Supplier:  LGC GENOMICS LLC 600521
Encompass
Lucigens E. cloni competent cells share the most useful genetic elements of standard cloning strains like DH5™, DH10B™, JM109, TOP10, etc. and directly replace them in cloning protocols. However, E. cloni electrocompetent cells incorporate a unique manufacturing technology that increases transformation efficiency, recombinant yields and reliability, while decreasing costs. These cells provide solutions for a wide range of applications at economical prices. E. cloni 10G & 10GF ELITE Electrocompetent Cells > 2 10^10 cfu/g pUC DNA. ELITE Cells have twice the transformation efficiency compared to ultra high efficiency cells from other suppliers. ELITE Cells provide large numbers of transformants from hard-to-clone fragments or limited DNA at a lower price.
Catalog No. NC0622723