Learn More
ATCC Escherichia coli (Migula) Castellani and Chalmers, Strain C36 - Freeze-Dried Research Strain (ATCC 8677)
Supplier: ATCC 8677
Escherichia coli (Migula) Castellani and Chalmers (ATCC 8677) is a freeze-dried bacterial strain supplied for laboratory research use. The strain is commonly employed as a bacteriophage host and can be revived and cultured on standard bacteriological media at 37°C. Supplied lyophilized for long-term stability; intended for research use only and handled according to institutional biosafety guidelines.
- Freeze-dried format for extended shelf life.
- Strain designation: C36.
- Organism: Escherichia coli (Migula) Castellani and Chalmers.
- Recommended growth temperature: 37°C.
- Recommended media: nutrient agar, nutrient broth, or tryptic soy broth.
- Storage conditions: 2°C to 8°C for recommended storage.
- Biosafety level: BSL 1; follow institutional safety procedures.
- Intended use: research use only; suitable as a bacteriophage host.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.