Learn More
Bon Opus Biosciences Recombinant E.coli 6-Phospho-Beta-Glucosidase/BGLA (N-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP053500UG
Encompass_Preferred
Recombinant E. coli BGLA is produced by E.coli. The target gene encoding M1-L479 is expressed with a 6His tag at the N terminus. Expression system: E. coli. Accession# Q46829. Formulation: Lyophilized frum a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# BP053
Catalog No. 50-208-0129