Learn More
Abcam Recombinant E. coli groEL protein, 500UG
Supplier:  Abcam AB51307500UG
Encompass_Preferred
Recombinant E. coli groEL protein is a Escherichia coli K-12 Full Length protein, in the 1 to 548 aa range, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: groL, mopA, b4143, JW4103, groEL, Chaperonin GroEL, 60 kDa chaperonin, Chaperonin-60, GroEL protein, Cpn60
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1948785