Learn More
Medchemexpress LLC (E)-Coniferyl alcohol | 32811-40-8 | 1 ML

Supplier: Medchemexpress LLC HYN4283A10MM/1ML
(E)-Coniferyl alcohol (CAS: 32811-40-8) is an isomer of Coniferyl alcohol. Coniferyl alcohol is an intermediate involved in the biosynthesis of eugenol, stilbenoids, and coumarin. This product specifically acts as a fungal inhibitor. It is provided as a 10 mM solution in 1 mL of DMSO.
- Isomer of coniferyl alcohol
- Inhibits fungal growth
- For research use only
- Purity of 99.62%
- White to off-white solid appearance
- Molecular weight of 180.20
- Chemical formula C10H12O3
- Classified as a phenol
- Sourced from plants like Rutaceae Maclurodendron oligophlebium (Merrill) T. G. Hartley and Salicaceae
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.