Learn More
Medchemexpress LLC (E)-Coniferyl alcohol | 32811-40-8 | 5 MG

Supplier: Medchemexpress LLC HYN4283A5MG
(E)-Coniferyl alcohol is an isomer of Coniferyl alcohol. Coniferyl alcohol acts as an intermediate in the biosynthesis of eugenol, stilbenoids, and coumarin. This compound specifically inhibits fungal growth. It is intended for research use only.
- Purity: 99.62%
- Appearance: Solid, white to off-white
- Molecular weight: 180.20
- Formula: C10H12O3
- Structure classification: Phenols
- Initial source: Plants Rutaceae Maclurodendron oligophlebium (Merrill) T. G. Hartley; Plants Salicaceae
- Solubility: DMSO: 125 mg/mL (693.67 mM; requires ultrasonic; hygroscopic DMSO significantly impacts solubility, use newly opened DMSO).
- Applications: Fungal inhibitor
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.