Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-coniferyl alcohol | 32811-40-8 | MFCD00002922 | 99.6% | 180.20 g/mol | C10H12O3 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN4283A10MG

Encompass_Preferred

(E)-Coniferyl alcohol is the E-isomer of coniferyl alcohol, a phenylpropanoid intermediate involved in plant secondary metabolism. It is supplied as a high-purity research reagent with reported antifungal activity and is intended for laboratory use in small-scale studies.

  • High purity suitable for analytical and biological experiments.
  • Antifungal activity reported in literature.
  • Solid, white to off-white appearance for easy handling.
  • Stable under recommended storage: sealed at 4°C; in solvent: -80°C (6 months), -20°C (1 month).
  • Supplied in milligram-scale packages for research use.
  • Molecular formula C10H12O3; molecular weight 180.20 g/mol.
  • CAS number 32811-40-8 for chemical identification.

Catalog No. 50-002-89800


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.