Learn More
ATCC E. Derm (NBL-6); Dermis; Horse (Equus caballus)
Supplier:  ATCC CCL57
Encompass
E. Derm (NBL-6) is a cell line exhibiting fibroblast morphology that was isolated in 1965 from the dermis of a 4-year-old, female quarter horse. This cell line was deposited by AJ Kniazeff, WA Nelson-Rees, and NB Darby. E. Derm (NBL-6).
Catalog No. 50-238-1811