Learn More
Medchemexpress LLC (E,E)-3,5-bis(4-phenyl-1,3-butadienyl)-4,4-difluoro-4-bora-3a,4a-diaza-s-indacene | 164106-16-5 | 99.2% | 448.31 g·mol⁻¹ | C29H23BF2N2 | 10 MG

Supplier: Medchemexpress LLC HYD145910MG
BODIPY 665/676 is a lipophilic, radical-sensitive fluorescent probe used to detect and study lipid peroxidation and radical-driven lipid autoxidation in biological samples. It is supplied as a dry dye for use in fluorescence microscopy and assays that monitor oxidative stress.
- Lipophilic, radical-sensitive fluorescent probe.
- Excitation/emission maxima at 665/676 nm.
- High purity (99.23%).
- Molecular weight 448.31 g·mol⁻¹; formula C29H23BF2N2.
- Suitable for fluorescence imaging and lipid peroxidation assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.