Learn More
Carbosynth LLC. H-Glu{Glu[Cyclo(Arg-Gly-Asp-D-Phe-Lys)]2}2, 5mg, MW: 2748.04, Molecular formula: C123H179N39O34
SDP
Supplier:  Carbosynth LLC. PCI3975PI5MG
Encompass_Preferred
Synonyms: H-Glu{Glu[cyclo(Arg-Gly-Asp-D-Phe-Lys)]2}22, E{E[cyclo(RGDfK)]2}2, H-E{E[c(RGDfK)]2}2, Application: Tetrameric RGD Peptide, Storage Temperature: -20°C, References: S. Liu, Z.J. He, W.Y. Hsieh, Y.S. Kim, and Y. Jiang, Bioconjugate Chem., 17, 1499 (2006)., S. Liu, W.Y. Hsieh, Y. Jiang, Y.S. Kim, S.G. Sreerama, X. Chen, B. Jia, and F. Wang,. Bioconjugate Chem., 18, 438 (2007).
Catalog No. 50-168-6360