Learn More
Medchemexpress LLC (E,E)-farnesol-d6 | 166447-71-8 | 98.6% | 228.40 | C15H20D6O | 1mg
SDP
Supplier:  Medchemexpress LLC HYY0248AS1MG
Encompass_Preferred
(E E) - Farnesol-d6 (trans trans-Farnesol-d6) is deuterium labeled (E E) - Farnesol (HY-Y0248) (E E)-Farnesol (trans trans-Farnesol) is a quorum-sensing molecule of Candida species (E E)-Farnesol can inhibit the growth metabolism and biofilm formation of various Candida species and affect their morphology and invasiveness[1][2]
Catalog No. 50-002-67939