Learn More
Medchemexpress LLC (E)-β-Farnesene (Standard) | 18794-84-8 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN7364R100MG
Encompass_Preferred
(E)-β-Farnesene (Standard) is the analytical standard of (E)-β-Farnesene. It is intended for research and analytical applications. (E)-β-Farnesene (trans-β-Farnesene) is a volatile sesquiterpene hydrocarbon found in *Phlomis aurea Decne* essential oil. It can be used as a feeding stimulant for the sand fly *Lutzomyia longipalpis*. This compound is an analytical standard, serving as a reference standard for assays. It is commonly used in qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS.
  • Analytical standard for (E)-β-Farnesene
  • Intended for research and analytical applications
  • Volatile sesquiterpene hydrocarbon
  • Can be used as a feeding stimulant for the sand fly *Lutzomyia longipalpis*
  • Reference standard for assays
  • Used in qualitative, quantitative, and methodological research experiments
  • Applications include HPLC, GC, and MS
Catalog No. 50-004-19380