Learn More
Medchemexpress LLC (E)-β-Farnesene (Standard) | 18794-84-8 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN7364R50MG
Encompass_Preferred
(E)-β-Farnesene (Standard) is the analytical standard of (E)-β-Farnesene, intended for research and analytical applications. It is a volatile sesquiterpene hydrocarbon found in Phlomis aurea Decne essential oil and can be used as a feeding stimulant for the sand fly Lutzomyia longipalpis.
  • Analytical standard for research and analytical applications
  • Volatile sesquiterpene hydrocarbon found in Phlomis aurea Decne essential oil
  • Can be used as a feeding stimulant for the sand fly Lutzomyia longipalpis
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-18014