Learn More
Medchemexpress LLC HY-N0060B 100mg Medchemexpress, (E)-Ferulic acid CAS:537-98-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0060B100MG
Encompass_Preferred
Medchemexpress, HY-N0060B 100mg (E)-Ferulic acid CAS:537-98-4 (E)-Ferulic acid is a isomer of Ferulic acid which is an aromatic compound, abundant in plant cell walls. (E)-Ferulic acid causes the phosphorylation of β-catenin, resulting in proteasomal degradation of β-catenin and increases the expression of pro-apoptotic factor Bax and decreases the expression of pro-survival factor survivin. (E)-Ferulic acid shows a potent ability to remove reactive oxygen species (ROS) and inhibits lipid peroxidation. (E)-Ferulic acid exerts both anti-proliferation and anti-migration effects in the human lung cancer cell line H1299. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0217