Learn More
Medchemexpress LLC (E)-ferulic acid-d3 | 860605-59-0 | 99.8% | 197.20 g/mol | C10H7D3O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0060BS10MG
Encompass_Preferred
(E)-Ferulic acid-d3 is the deuterium-labeled analogue of (E)-ferulic acid used as an analytical standard and tracer for research applications. It is intended for use as an internal standard in quantitative techniques and exhibits the antioxidant and anti-proliferative properties associated with the parent compound.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity (99.8%) suitable for analytical applications.
  • Soluble in DMSO at 100 mg/mL; ultrasonic treatment may be required.
  • Stable as a powder at -20°C for up to 3 years under recommended storage.
  • White to off-white solid appearance for easy visual inspection.
Catalog No. 50-002-96974