Learn More
Medchemexpress LLC (E)-Flavokawain A | 37951-13-6 | MFCD00017174 | 20 MG

Supplier: Medchemexpress LLC HYN510620MG
(E)-Flavokawain A is a chalcone extracted from Kava with significant anticarcinogenic effects. This compound has been shown to induce apoptosis in bladder cancer cells via a Bax protein-dependent and mitochondria-dependent pathway, effectively suppressing tumor growth in mice. It is intended for research use only.
- Chalcone extracted from Kava
- Exhibits anticarcinogenic effects
- Induces apoptosis in bladder cancer cells
- Suppresses tumor growth in mice
- Purity of 99.53%
- Molecular weight of 314.33
- Chemical formula C18H18O5
- Appears as a light yellow to yellow solid
- Store at 4°C, protected from light
- When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.